PYREX® Reusable Volumetric Pipettes, Corning
53043-012
: Corning
Product available on GSA Advantage®
- Capacidad:0.5 ml
- Descripción:PYREX® reusable volumetric pipet
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Referencia:53043-012
- Nº de proveedor:7102-1X
These reusable pipettes are designed primarily for schools and laboratories where the Class A tolerance is not required.
- Larger tip openings for faster delivery
- Color-coded
- Colored graduations
- Accuracy tolerances are twice those specified for Class A volumetric ware
They are constructed to the same wall thickness and length as standard Class A volumetric pipettes, but have larger tip openings for faster delivery. Primarily due to the faster delivery, accuracy tolerances are twice those specified for Class A volumetric ware. Reference: ASTM E-969.