PYREX® Distilling Adapter, Corning
PYREX® Distilling Adapter
Supplier: Corning
Número de proveedor #: 8950-24
PYREX® Distilling Adapter, Corning
Catalog # 62939-996
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
These PYREX® 60° angle distilling connecting adapters have a 24/40 Standard Taper outer joint at the upper end and inner 24/40 Standard Taper joints and on the sidearm and lower joints
This tube also serves as a replacement part for organic chemistry kit No. 6949
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:PYREX® Distilling Adapter
- Cat. No.:62939-996
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact