Orders LinkContactUsLinkComponent
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate Disinfectant, Contec®
Supplier:  CONTEC, INC. MX
Certificates
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate Disinfectant, Contec®
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PREempt™ Concentrate Disinfectant Solution works fast for large areas to ensure user, protocol, and product compliance with fast contact times and broad-spectrum efficacy
  • 3 minute Broad-Spectrum Sanitizing (1:128)
  • 30 second Broad-Spectrum Sanitizing (1:16)
PREempt™ Concentrate utilizes AHP®, a patented synergistic blend of commonly used and safe ingredients which, when combined with low levels of hydrogen peroxide, dramatically increase the potency and cleaning performance.
The concentrated formula requires dilution and is cost effective for large surfaces.
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 10822-520
    PREempt™ Concentrate
    PREempt™ Concentrate
    4×3.8 L (1 gal.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (4UN)
    Case (4UN) - $5,453.75
    $5,453.75
    Case (4UN)
       $5,453.75
    Supplier Number: 21305
  • 10822-522
    PREempt™ Concentrate
    PREempt™ Concentrate
    200 L (55 gal.)
    Discontinued
    Supplier Number: -
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 