Orders LinkContactUsLinkComponent
PYREX® Ignition Tube, Corning
PYREX® Ignition Tube, Corning
   Opciones sostenibles disponibles
Supplier:   Corning
PYREX® Ignition Tube, Corning
Supplier:  Corning
   Opciones sostenibles disponibles
Certificates
PYREX® Ignition Tube, Corning
3 Options Available Below
Product Details & Documents
These PYREX® rimless ignition tubes are made from No. 7740 glass.
  • Very good chemical resistance
  • Good temperature resistance
These ignition tubes are satisfactory for most applications encountered in school work where temperatures above 600 °C are not required. These rimless tubes perform well when continually used at temperatures up to 500 °C, or for short periods to 600 °C.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 89091-156
  • 89091-184
  • 60835-247
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Especificaciones
Product Family Options
Información del producto
  • Material
    Dimensions
    Availability
    Price
  • 89091-156
    Ignition Test Tube
    Ignition Test Tube
    PYREX® glass
    14 x 100 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (288UN)
    Case (288UN) - $32,890.00
    $32,890.00
    Case (288UN)
       $32,890.00
    Supplier Number: 9860-14
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-156
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 89091-184
    Ignition Test Tube
    Ignition Test Tube
    PYREX® glass
    16 x 125 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (288UN)
    Case (288UN) - $27,460.00
    $27,460.00
    Case (288UN)
       $27,460.00
    Supplier Number: 9860-16
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89091-184
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 60835-247
    Ignition Test Tube
    Ignition Test Tube
    PYREX® glass
    25 x 200 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 24
    Paquete de 24 - $5,270.00
    $5,270.00
    Log in Para disponibilidad
    Case de 5 (24UN)
    Case de 5 (24UN) - $26,294.22
    $26,294.22
    Paquete de 24
       $5,270.00
    Case de 5 (24UN)
       $26,294.22
    Supplier Number: 9860-25
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    60835-247
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact