HPLC Columns, YMC-Pack ODS-A
Product Details & Documents
ODS-A is a high coverage, fully endcapped C18 phase which exhibits exceptional batch-to-batch reproducibility.
- Highly versatile ODS phase
- Selectivity for polar to moderately nonpolar pharmaceuticals, organic chemicals, biologicals and natural products
ODS-A is the first choice of many chromatographers for high performance reversed phase separations. It has selectivity for the separation of a wide range of organic compounds found in pharmaceutical, chemical and biotechnology industries. ODS-A is a high coverage, fully endcapped C18 phase which exhibits exceptional batch-to-batch reproducibility. ODS-A columns are noted for their high efficiency, selectivity and long lifetimes.
These are noted for their high efficiency, selectivity and long lifetimes.
Especificaciones
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Pore size:120 Å
- Length:250 mm
- Phase:ODS-A
- Carbon load:0.17
- Cat. No.:97028-150
- Endcapped:Yes
- USP code:L1
- pH range:2.0 - 7.5
- Surface area:175 m2/g