Rxi®-624Sil MS Columns (Fused Silica), Restek
Supplier: RESTEK CORP
Certificates
About this item
Rxi® fused silica capillary GC columns are designed specifically for exceptional inertness, ultra-low bleed, and reproducible retention and selectivity.
- Low-bleed, high-thermal stability column—maximum temperatures up to 300 to 320 °C
- Inert—excellent peak shape for a wide range of compounds
- Selective—G43 phase highly selective for volatile organics and residual solvents, great choice for USP 467
- Manufactured for column-to-column reproducibility—well-suited for validated methods
- Mid-polarity Crossbond® phase
In addition to outstanding performance, this combination of column characteristics assures Rxi® columns will have long lifetimes. The Restek chemists also have developed a stable, reproducible manufacturing process, so that Rxi® columns will meet your needs every time you install a new column.
Specifications
- Catalog No:
- 10058-472
- 10058-470
- Descripción:
- Rxi-624Sil MS
- Rxi-624Sil MS
- Temperature limit:
- -20 to 300/320 °C
- -20 to 300/320 °C