ContactUsLinkComponent
Polycarbonate Face Shields, Oberon
Polycarbonate Face Shields, Oberon
Catalog # 87001-674
Supplier:  SUREWERX USA INC
undefined
Polycarbonate Face Shields, Oberon
Catalog # 87001-674
Supplier:  SUREWERX USA INC
Supplier Number:  179R
Specifications
  • Descripción:
    Face Shield with Ratchet Head Gear
  • Referencia:
    87001-674
  • Nº de proveedor:
    179R
Specifications
About this item
The clear UV Protective Series face shields provide 100% protection from UV energy from 200 to 400nm.
  • Scratch resitant outside
  • Antifog inside
Not suitable for electrical arc flash protection, not intended for high intensity laser type applications.
Certifications: Meets ANSI Z87.1-2010 standards.