PYREX® Volumetric Pipettes, Class A, Serialized, Corning
Supplier: Corning
Certificates
About this item
These PYREX® volumetric pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
- Reusable precision pipettes
- Color-coded markings are enameled onto the glass
PYREX® serialized/certified volumetric glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Each pipette is individually serialized and supplied with a Certificate of Identification and capacity, traceable to NIST standards. These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line) and 'To Deliver' (TD: top line) to specifications for Class A volumetric ware.
Specifications
- Catalog No:
- 46610-116
- 46610-118
- 46610-108
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Related Information
Frequently Bought Together