ContactUsLinkComponent
PYREX® Media Storage Bottle with High Temperature Cap, Corning
PYREX® Media Storage Bottle with High Temperature Cap, Corning
Catalog # 10416-312
Supplier:  Corning
undefined
PYREX® Media Storage Bottle with High Temperature Cap, Corning
Catalog # 10416-312
Supplier:  Corning
Supplier Number:  10416-312
Specifications
  • Tamaño de la tapa:
    GL 45
  • Intervalo de graduación:
    100 ml
  • Rango de graduación:
    100 – 900 mL
  • Capacidad:
    1 L
  • Descripción:
    PYREX® Storage Bottle
  • Altura:
    225 mm
  • Referencia:
    10416-312
  • Neck O.D.:
    101 mm
  • Nº de proveedor:
    1395-1LHTC
Specifications
About this item
Heavy duty bottles can be used for storage, mixing, and sampling.
Bottles have permanent white enamel graduations and a marking spot.
Ordering information: Includes red PBT cap and ETFE pouring ring for dry heat sterilization.