ContactUsLinkComponent
Detection of GMOs Kit
Detection of GMOs Kit
Catálogo # 470136-468
Proveedor:  EDVOTEK, INC.
undefined
Detection of GMOs Kit
Catálogo # 470136-468
Proveedor:  EDVOTEK, INC.
Número de proveedor:  121
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Especificaciones
  • Descripción:
    Detection of GMOs Kit
  • Referencia:
    470136-468
  • Nº de proveedor:
    121
Especificaciones
Acerca de este artículo
For centuries, humans have used selective breeding and conventional hybridization to produce desirable qualities and to increase crop yields. Today, scientists use genetic engineering to directly manipulate the DNA, quickly producing these desirable traits. In this experiment, students will use agarose gel electrophoresis to explore the molecular methods used by scientists to identify genetically modified organisms. No thermal cycler is required. Students are also encouraged to explore the controversy surrounding the use of genetically modified organisms.