ContactUsLinkComponent
Exploring the Bacteriophage Life Cycle
  470136-510
undefined
Exploring the Bacteriophage Life Cycle
  470136-510
 :  209
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Descripción:
    Exploring the Bacteriophage Life Cycle
  • Referencia:
    470136-510
  • Nº de proveedor:
    209
Although bacterial viruses, or bacteriophages, are present in many natural environments, they cannot survive autonomously. They require a host cell to reproduce and survive. In this experiment, students will learn about the different life cycles of bacterial viruses. They will then perform a viral plaque assay to indirectly visualize the viruses and to determine viral titer and multiplicity of infection (MOI).
Note: If shipping to warehouse, perishable coupon included.