Glass Microscope Slides
Glass slides, 75×25 mm (3×1")
Catalog # 470235-792
Supplier: GSC INTERNATIONAL SE
Número de proveedor #: 4-13051-25
Jump to:
Product Details & Documents
Make your own Slides with these High Grade Glass Slides
- Quality slide making supplies
- Traditional slide type
These slides are specially developed for our labs and processed with the utmost care to provide unmatched optical clarity, cleanliness, and exact tolerances.
Especificaciones
- Description:Glass slides
- L×W:75×25 mm (3×1")