Orders LinkContactUsLinkComponent
ESD Two-Layer Rubber Table Matting, Transforming Technologies
ESD Two-Layer Rubber Table Matting, Transforming Technologies
ESD Two-Layer Rubber Table Matting, Transforming Technologies
Certificates
ESD Two-Layer Rubber Table Matting, Transforming Technologies
ESD Two-Layer Rubber Table Matting, Transforming Technologies
2 Options Available Below
Product Details & Documents
Anti-static workstation mats are made of a durable two-ply rubber that offers excellent resistance to hot solders, soldering irons, and most solvents. A conductive layer backs the static dissipative top layer, creating outstanding electrical performance characteristics. Available in Nasa Blue, Royal Blue, Green, or Gray with the following features:
Dissipative work surface: 10E6 – 10E8 ohms Rtt
Thickness: 0.060" or 0.080"
Resistance to Ground: 8 x 10E6-8 ohms
Resistance Point to Point: 2 x 10E6-8 ohms
Product Family Options
ESD Table Mat Rolls
Información del producto
  • Color
    Thickness
    Size
    Availability
    Price
  • Gray
    0.060"
    61 cm x 15.2 m (24" x 50')
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $11,090.00
    $11,090.00
    Each (1UN)
       $11,090.00
    Supplier Number: MT2524GY
  • Green
    0.080"
    91.4 cm x 15.2 m (36" x 50')
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $17,890.00
    $17,890.00
    Each (1UN)
       $17,890.00
    Supplier Number: MT4536GN