Orders LinkContactUsLinkComponent
Tedlar® Sampling Bags, Restek
Supplier:   RESTEK CORP
Tedlar® Sampling Bags, Restek
Supplier:  RESTEK CORP
Certificates
Tedlar® Sampling Bags, Restek
Tedlar® Sampling Bags, Restek
2 Options Available Below
Product Details & Documents
Sampling bags are a low-cost whole-air sampling device for high-level VOCs and permanent gases.
  • Find the bags you need - we offer sizes from 0.5 to 100 L
  • Unique all-in-one septum and valve fitting make these lightweight and easy to use
  • Polypropylene valve conveniently connects to ³/₁₆" Int.Ø PTFE tubing
  • Continuous sampling temperature up to 107 °C (225 °F); short term (1 to 2 hours) temperature up to 176 °C (350 °F)
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplace atmospheres, ambient, indoor air quality, and breath analysis. Choose the film type appropriate for your application. All Restek's bags feature a polypropylene combo valve with hose connection to fit ³/₁₆" Int.Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
Product Family Options
Product Information
  • Capacity
    W×L
    Availability
    Price
  • 10803-272
    Tedlar bag with PP valve and septum fitting
    Tedlar bag with PP valve and septum fitting
    0.5 L
    6×6"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $6,743.14
    $6,743.14
    Paquete de 10
       $6,743.14
    Supplier Number: 22049
  • 10058-714
    Tedlar bag with PP valve and septum fitting
    Tedlar bag with PP valve and septum fitting
    1 L
    7×7"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $7,360.94
    $7,360.94
    Paquete de 10
       $7,360.94
    Supplier Number: 22050