Orders LinkContactUsLinkComponent
Avian Development Slides
Supplier:   Avantor
Avian Development Slides
Supplier:  Avantor
Certificates
Avian Development Slides
Avian Development Slides
6 Options Available Below
Product Details & Documents
Explore Developmental Biology.
  • Investigate varying ages of developing chicks
  • Illustrates specific structures at developmental stages
  • Clearly distinguishable features
Chickens are historic model organisms for developmental studies. They are particularly useful in comparative anatomy and vertebrate embryology studies because of the similarities in embryological development across many vertebrate species, including humans.
 : Choose from a variety of individual slides ranging from 12-92 hr. development in an assortment of slide types.
Product Family Options
Información del producto
  • Orientation
    Stain
    Availability
    Price
  • 470182-468
    Chick, 40-Hour
    Chick, 40-Hour
    Whole mount
    Schneider's
    Discontinued
  • 470177-030
    Chick, 22 – 30 Hours
    Chick, 22 – 30 Hours
    Whole mount
    Schneider's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $740.40
    $740.40
    Each (1UN)
       $740.40
  • 470177-032
    Chick, 46 – 56 Hours
    Chick, 46 – 56 Hours
    Whole mount
    Schneider's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $468.50
    $468.50
    Each (1UN)
       $468.50
  • 470182-488
    Chick, 57 – 72 Hours
    Chick, 57 – 72 Hours
    Whole mount
    Schneider's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $708.60
    $708.60
    Each (1UN)
       $708.60
  • 470177-034
    Chick, 73 – 96 Hours
    Chick, 73 – 96 Hours
    Whole mount
    Schneider's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $493.15
    $493.15
    Each (1UN)
       $493.15
  • 470182-438
    Chick, 12 – 21 Hours
    Chick, 12 – 21 Hours
    Whole mount
    Schneider's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $441.00
    $441.00
    Each (1UN)
       $441.00
  • 470182-442
    Chick, 18-Hour
    Chick, 18-Hour
    Whole mount
    Schneider's
    Discontinued
  • 470182-460
    Chick, 31 – 45 Hours
    Chick, 31 – 45 Hours
    Whole mount
    Schneider's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $729.00
    $729.00
    Each (1UN)
       $729.00