Orders LinkContactUsLinkComponent
Banana Plug Wires
Banana Plug Wires
Certificates
Banana Plug Wires
Banana Plug Wires
9 Options Available Below
Product Details & Documents
Banana Plug Wires are a learn-by-building introduction to a motor’s basic components.
  • Two-Way
  • Stackable
  • Size: 4 mm
  • Various Lengths Available
  • Flexible, 18-Gauge Stranded Wire Cord
A flexible, 18-gauge stranded wire cord has two-way, stackable, 4 mm banana plugs at both ends.
Banana plugs available in various lengths and pack sizes.
Product Family Options
Información del producto
  • Color
    Length
    Availability
    Price
  • 470219-738
    Banana plug wire pack of 6
    Banana plug wire pack of 6
    12"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 6
    Paquete de 6 - $438.14
    $438.14
    Paquete de 6
       $438.14
    Supplier Number: WBP012-PK/6
  • 470219-740
    Banana plug wire pack of 6
    Banana plug wire pack of 6
    24"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 6
    Paquete de 6 - $431.75
    $431.75
    Paquete de 6
       $431.75
    Supplier Number: WBP024-PK/6
  • 470219-742
    Banana plug wire pack of 6
    Banana plug wire pack of 6
    36"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 6
    Paquete de 6 - $534.08
    $534.08
    Paquete de 6
       $534.08
    Supplier Number: WBP036-PK/6
  • 470219-744
    Banana plug wire
    Banana plug wire
    Black
    12"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $60.21
    $60.21
    Each (1UN)
       $60.21
    Supplier Number: WBP012-R
  • 470219-746
    Banana plug wire
    Banana plug wire
    Black
    12"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $60.21
    $60.21
    Each (1UN)
       $60.21
    Supplier Number: WBP012-B
  • 470219-748
    Banana plug wire
    Banana plug wire
    Red
    24"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $63.96
    $63.96
    Each (1UN)
       $63.96
    Supplier Number: WBP024-R
  • 470219-750
    Banana plug wire
    Banana plug wire
    Black
    24"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $63.96
    $63.96
    Each (1UN)
       $63.96
    Supplier Number: WBP024-B
  • 470219-752
    Banana plug wire
    Banana plug wire
    Red
    36"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2.00
    $2.00
    Liquidación
    Each (1UN)
       $2.00
    Supplier Number: WBP036-R
  • 470219-754
    Banana plug wire
    Banana plug wire
    Black
    36"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2.00
    $2.00
    Liquidación
    Each (1UN)
       $2.00
    Supplier Number: WBP036-B