Convection Apparatus
Catalog # 470219-718
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Convection Apparatus
- Referencia:470219-718
- Nº de proveedor:CGA001
Specifications
About this item
This apparatus consists of a metal box (22 x 10 x 11cm) with clear acrylic front panel, two glass chimneys, a candle, and a cotton filled wick as the smoke source. Includes activity guide.
- Clear panel allows the flow to be seen easily
Shipping weight: 1.25 lbs.