Knife-Edge Clamp
Knife-Edge Clamp
470219-710
: CLMKE1
This clamp can be attached to any standard meter stick. It is used as a knife-edge fulcrum for supporting the meter stick in the inverted position.
- Fits standard meter sticks
- Thumbscrew to hold securely
The knife edge clamp is useful when invetsigating torque on a lever.
- Description:Knife-Edge Clamp