Orders LinkContactUsLinkComponent
Complete Meter Stick Optics Bench
Complete Meter Stick Optics Bench
Complete Meter Stick Optics Bench
Certificates
Complete Meter Stick Optics Bench
Complete Meter Stick Optics Bench
2 Options Available Below
Product Details & Documents
Classical experiment for studying optics on a budget.
  • All components connect to any standard meter stick
Student optical bench contains sufficient equipment for high school physics applications.
Contents:
1 Meter Stick
1 Screen support
2 lens supports for 38mm and 75mm lenses/mirrors
5 Board screens measuring 10 x 125 cm with a millimeter scale on both sides
1 Marker and object
2 Metal supports for meter stick
1 Candle holder
1 Paraffin candle
1 Activity guide
Required but not included are lenses. Focal lengths under 50cm work best.
Product Family Options
Información del producto
  • No. of Lab Groups
    Availability
    Price
  • 470219-700
    Complete Meter Stick Optics Bench
    Complete Meter Stick Optics Bench
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $326.74
    $326.74
    Each (1UN)
       $326.74
    Supplier Number: OBSET1
  • 470219-702
    Meter Stick Optics Bench, No Meter Stick
    Meter Stick Optics Bench, No Meter Stick
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $207.34
    $207.34
    Each (1UN)
       $207.34
    Supplier Number: OBSET2-NS