Orders LinkContactUsLinkComponent
Ward's® Plastic Slide Boxes
Ward's® Plastic Slide Boxes
Supplier:   VWR International
Ward's® Plastic Slide Boxes
Supplier:  VWR International
Certificates
Ward's® Plastic Slide Boxes
Ward's® Plastic Slide Boxes
2 Options Available Below
Product Details & Documents
The plastic slide box protects 1x3" slides and won't warp when temperature and humidity vary.
  • Plastic construction
  • Foam-lined bottom
  • Numbered insert for easy organization of slides
The 25-capacity and 100-capacity boxes have fully removable lid latches with a plastic clip. The 10-capacity boxes are hinged, with a snap-close mechanism.
Product Family Options
Product Information
  • Availability
    Price
  • 470151-632
    25-Capacity Slide Box
    25-Capacity Slide Box
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $58.58
    $58.58
    Log in for availability
    Case de 12 (1UN)
    Case de 12 (1UN) - $63.85
    $63.85
    Each (1UN)
       $58.58
    Case de 12 (1UN)
       $63.85
  • 470151-630
    100-Capacity Slide Box
    100-Capacity Slide Box
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $182.18
    $182.18
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $198.58
    $198.58
    Each (1UN)
       $182.18
    Case de 6 (1UN)
       $198.58