Pro-Wipe® 750 Polypropylene/Cellulose Composite Nonwoven Cleanroom Wipes, Berkshire
Catalog # 21914-408
Supplier: BERKSHIRE CORP
Small Business Enterprise
Specifications
- Esterilidad:Non sterile
- Matriz:PP/cellulose
- Fibras (>100 µm):7.1/cm²
- Peso base:65 g/m²
- Absorbancia:424 ml/m²
- Absorbancia, velocidad:2 s
- Referencia:21914-408
- Empaquetado:75/pack, 20 packs/case
- An x La:30.5×30.5 cm (12×12")
- Nº de proveedor:PW750121220
Specifications
About this item
Pro-Wipe® 750 is a solvent-resistant wipe with exceptional absorbency that makes an excellent upgrade for industrial wipe users. It is composed of a layer of cellulose bonded between two outer layers of polypropylene.
- Excellent combination of polyester strength and cleanliness with the absorbent characteristics of cellulose
- Nonwoven wipes with knife-cut edge
- Smooth, highly absorbent, and durable
- Low extractable levels and particle counts
- Chemically compatible with common cleaning and disinfecting solutions
These wipes are engineered for compatibility with solvents, and are specifically designed for the removal of etchants and other chemical spills. The wipes are chemically compatible with common cleaning and disinfecting solutions. High absorbency to weight ratio may reduce wipe usage and landfill waste pounds compared to similar weight. They are excellent for general wiping and cleaning in biomedical environments.
Certifications: Designed for use in ISO class 5–8. Cleanroom class 100 – 100,000. EC regulation 1935 (2004)- Food contact compliant. FDA 21 CFR -Food contact compliant. USP <797> applications.