Polyurethane Foam Sock Swabs, Berkshire
Catalog # 10810-317
Supplier: BERKSHIRE CORPORATION MX
Small Business Enterprise
Specifications
- Descripción:BCR® Large Open Cell Foam Swab
- Longitud:27 mm (5")
- Referencia:10810-317
- Tamaño de la punta:12.7 mm (1/2")
- Nº de proveedor:LT000125.10
- Handle Type:Polypropylene
Specifications
About this item
These general purpose swabs are designed for cleaning small and confined areas.
- Low particles, fibers, ions, and extractables
- Good chemical resistance
- Soft and non-abrasive with no contaminating adhesives
- Good sorbency
- Clean room class 100 to 100000
Its laundered open cell foam head combines superior cleanliness with particle entrapment capability.
Ordering information: 100 swabs per pack, 10 packs per case (double packed).