About this item
The VWR® Water Jacketed CO2 incubator protects your cultures from unwanted heat loss by surrounding them with triple-walled construction and a jacket of temperature-conditioned water.
- 6.5 cu. ft. internal chamber volume
- Supplied with four removable stainless steel shelves and water pan
- Outstanding temperature stability and security
- Fan-assisted airflow and in-chamber sensors maintain a uniform and stable growth environment
- Choice of long-lasting TC or independent IR CO2 sensors
- Dual temperatures for over-temperature protection
- Easy assembly and disassembly of interior without tools
- Dual stack configurations include mobile support stand and stacking kit to maximize lab space
- 3-Year Part and Labor Warranty
An ideal choice for laboratories concerned with temperature fluctuations from power outages or significant ambient temperature fluctuations.
In-chamber HEPA filtration airflow is designed to protect samples from airborne contaminants during routine access.
Feel safe with a VWR® CO2 incubator that provides you with selection, performance and quality needed in today’s cell culture laboratories. VWR® family of CO2 incubators include water jacketed and air jacketed configurations to meet specific culturing preferences and a wide range of solutions to meet capacity and budget needs. A choice of platform designs and capacities are available to meet your specific culturing preferences. Options include capacities ranging from 1.4 to 6.5 cu. ft., water jacketed models for outstanding temperature stability, and low maintenance air jacketed models with high temperature decontamination cycles. Choice of small capacity models for low volume culturing needs. Stackable models are available to maximize lab space. Choose between thermal conductivity or infrared CO2 sensors to meet preference and application needs. Optional available enhanced contamination control technologies include HEPA filtration and high temperature decontamination. These advanced technologies and contamination control provide a safe growth environment for your cultures. Feel confident that your cultures are in a stable and protected growth environment. Exceptional value for your lab. Fan assisted airflow designed for rapid recovery and uniform conditions. In-chamber probes and sensors designed for precise control. Dual temperatures for over-temperature protection. Polished, high grade stainless steel chambers.
Specifications
- Catalog No:
- 10810-744
- Detalles eléctricos:
- 120V