Equivalence C18, HPLC Columns
Proveedor: Avantor
Certificados
Acerca de este artículo
Equivalence C18 columns have been specifically developed as a high quality, cost effective replacement for many HPLC column brands.
- Excellent efficiency and reproducibility
- Available in 3 or 5 µm particle sizes
Maximum recommended pH: 2 to 8.
Especificaciones
- Catalog No:
- 10829-198
- Environmentally Preferable:
- Low Manufacturing Impact
- Certificación ambiental:
- ISO 14001:2015 (530840 UM15)
Especificaciones
Información relacionada
Certificados
Buscar por número de loteAYN4_Analytical_Chemistry_-_Chromatography Hichrom ACE Equivalence C18 HPLC Columns Brochure
Documentación del producto
Frequently Bought Together