ContactUsLinkComponent
Acrylic Pipe Demonstration
Catalog # 470219-530
undefined
Acrylic Pipe Demonstration
Catalog # 470219-530
Supplier Number:  PH100LCB
Specifications
  • Descripción:
    Acrylic Pipe Demonstration
  • Referencia:
    470219-530
  • Nº de proveedor:
    PH100LCB
Specifications
About this item
Show students how light reflects inside an acrylic pipe.
  • Transmits light like fiber optic cable
This hard acrylic 1/2" diameter light pipe is 2 feet of pipe coiled up at one end to demonstrate light transmission end to end. This light pipe is approx. 9" wide due to the coil in it.