CENCO® Spectrometer Prisms
Supplier: EISCO SCIENTIFIC LLC
Jump to:
About this item
Spectrometer prisms offer a different index of refraction for every need.
- Different Indices of Refractions for Every Need
- Glass Material
- Designed to Yield Accurate Results
Although any prism can be used in a spectrometer, these are specially designed to yield accurate results for spectroscopy experiments.
Choose your prism type and the approximate index of refraction that would work best for your experiment.
5 Options Available Below
Product Details & Documents
Spectrometer prisms offer a different index of refraction for every need.
- Different Indices of Refractions for Every Need
- Glass Material
- Designed to Yield Accurate Results
Although any prism can be used in a spectrometer, these are specially designed to yield accurate results for spectroscopy experiments.
Choose your prism type and the approximate index of refraction that would work best for your experiment.
Product Family Options
Product Information
- AvailabilityPrice
- 470219-498Prism Borosilicate Crown Glass, 1.51Supplier Number: PH0583A
- 470219-500Prism Dense Flint Glass, 1.62Supplier Number: PH0583B
- 470219-502Prism Flint Glass, 1.65Supplier Number: PH0583C
- 470219-504Prism Double Flint, 1.71-1.74Supplier Number: PH0583D
- 470219-506Prism Equilateral Flint, 1.62Supplier Number: PH0583E