Orders LinkContactUsLinkComponent
Optical Refraction Components
Optical Refraction Components
Optical Refraction Components
Optical Refraction Components
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
  • Students Measure the Refraction of Light
  • Refraction Blocks, Prisms, and Lenses
  • Optically Worked Surfaces
  • Bubble-Free Glass
  • Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
Product Family Options
Block
  • Description
    Material
    Dimensions
  • Rectangular Block
    Glass
    75 x 50 x 12 mm
    Each (1UN)
    Each (1UN) - $2.10
    $2.10
    Each (1UN)
       $2.10
     : PH0566A
  • Rectangular Block
    Glass
    75 x 50 x 18 mm
    Each (1UN)
    Each (1UN) - $194.02
    $194.02
    Each (1UN)
       $194.02
     : PH0566B
  • Rectangular Block
    Glass
    100 x 60 x 18 mm
    Each (1UN)
    Each (1UN) - $296.89
    $296.89
    Each (1UN)
       $296.89
     : PH0566C
  • Rectangular Block
    Glass
    125 x 65 x 18 mm
    Each (1UN)
    Each (1UN) - $378.97
    $378.97
    Each (1UN)
       $378.97
     : PH0566E
  • Rectangular Block
    Acrylic
    75 x 50 x 18 mm
    Each (1UN)
    Each (1UN) - $231.34
    $231.34
    Each (1UN)
       $231.34
     : PH0568A
  • Rectangular Block
    Acrylic
    100 x 75 x 18 mm
    Each (1UN)
    Each (1UN) - $301.67
    $301.67
    Each (1UN)
       $301.67
     : PH0568B
  • Rectangular Block
    Acrylic
    115 x 65 x 18 mm
    Each (1UN)
    Each (1UN) - $301.67
    $301.67
    Each (1UN)
       $301.67
     : PH0568C
  • Rectangular Block
    Acrylic
    125 x 65 x 18 mm
    Each (1UN)
    Each (1UN) - $329.92
    $329.92
    Each (1UN)
       $329.92
     : PH0568D
Prism
  • Description
    Material
    Dimensions
  • Equilateral Prism
    Acrylic
    75 sides x 9 mm Thick
    Each (1UN)
    Each (1UN) - $162.56
    $162.56
    Each (1UN)
       $162.56
     : PH0569A
  • Equilateral Prism
    Glass
    75 sides x 9 mm Thick
    Each (1UN)
    Each (1UN) - $231.34
    $231.34
    Each (1UN)
       $231.34
     : PH0569B
Semicircular
  • Description
    Material
    Dimensions
  • Semi Circular Block
    Acrylic
    90 x 16 mm
    Each (1UN)
    Each (1UN) - $238.80
    $238.80
    Each (1UN)
       $238.80
     : PH0570A
  • Semi Circular Block
    Acrylic
    90 x 25 mm
    Each (1UN)
    Each (1UN) - $315.53
    $315.53
    Each (1UN)
       $315.53
     : PH0570B
  • Semi Circular Block
    Glass
    90 x 16 mm
    Each (1UN)
    Each (1UN) - $4.95
    $4.95
    Each (1UN)
       $4.95
     : PH0570C
  • Semi Circular Block
    Glass
    90 x 18 mm
    Each (1UN)
    Each (1UN) - $265.44
    $265.44
    Each (1UN)
       $265.44
     : PH0570D
  • Semi Circular Block
    Glass
    100 x 18 mm
    Each (1UN)
    Each (1UN) - $287.28
    $287.28
    Each (1UN)
       $287.28
     : PH0570E
Frosted Lens
  • Description
    Material
    Dimensions
  • Double Convex
    Acrylic, frosted
    200 mm Focal Length
    Each (1UN)
    Each (1UN) - $411.48
    $411.48
    Each (1UN)
       $411.48
     : PH0577A
  • Double Concave
    Acrylic, frosted
    –200 mm Focal Length
    Each (1UN)
    Each (1UN) - $491.96
    $491.96
    Each (1UN)
       $491.96
     : PH0577B