Orders LinkContactUsLinkComponent
Optical Prisms
Optical Prisms
Optical Prisms
Certificates
Optical Prisms
8 Options Available Below
Product Details & Documents
Right angle and equilateral prisms are available in a variety of forms.
  • For Investigating Refraction and Creating Rainbows
  • Non-Optically Worked
  • Polished Faces
  • Beveled Edges
  • Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
Product Family Options
Right Angled Prisms
Información del producto
  • Material
    Size
    Availability
    Price
  • Glass
    25 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3.50
    $3.50
    Liquidación
    Each (1UN)
       $3.50
    Supplier Number: PH0558A
  • Glass
    38 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $165.22
    $165.22
    Each (1UN)
       $165.22
    Supplier Number: PH0558B
  • Glass
    50 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $7.40
    $7.40
    Liquidación
    Each (1UN)
       $7.40
    Supplier Number: PH0558C
  • Acrylic
    35 mm, 25 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $133.26
    $133.26
    Each (1UN)
       $133.26
    Supplier Number: PH0562A
  • Acrylic
    55 mm, 38 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $224.93
    $224.93
    Each (1UN)
       $224.93
    Supplier Number: PH0562B
  • Acrylic
    74 mm, 50 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $357.65
    $357.65
    Each (1UN)
       $357.65
    Supplier Number: PH0562C
Equilateral Prisms
Información del producto
  • Material
    Size
    Availability
    Price
  • Acrylic
    50 mm, 50 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $7.40
    $7.40
    Liquidación
    Each (1UN)
       $7.40
    Supplier Number: PH0560B
  • Acrylic
    63 mm, 63 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $505.83
    $505.83
    Each (1UN)
       $505.83
    Supplier Number: PH0560C