Universal Mirror
Universal stainless steel mirror, one side flat and one side curved, 3.5", 0.75"
Catalog # 470219-444
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PH0528
Jump to:
Product Details & Documents
With this stainless steel mirror, students can perform experiments using light rays with plane, concave, and convex mirrors all in one convenient and easy to use tool. The flexible metal curve can be switched to convert from a concave to convex mirror.
Especificaciones
- Description:Universal stainless steel mirror, one side flat and one side curved
- Height:0.75"
- Length:3.5"