Orders LinkContactUsLinkComponent
Compound Bar
Compound Bar; Brass and Steel
  470219-434
 :  PH0384B
Compound Bar
  470219-434
Compound Bar
Compound Bar
Compound Bar; Brass and Steel
$329.40
Each (1UN)
For demonstrating the differential expansion of two metals as shown by the curvature produced when the bar is heated. Each metal band is 200 x 15mm and riveted together.
  • Description:
    Compound Bar; Brass and Steel