Spring Large - Compression
Spring Large - Compression
470219-412
Número de proveedor #: PH0115C
Excellent demonstration of compression for hooke's law. Approximately 300mm long with a 10mm diameter and wire of 19 SWG.
- Description:Spring Large - Compression