SP Bel-Art Disposal Carton for Glass, Bel-Art Products, a part of SP
Catalog # 30617-648
Supplier: Bel-Art Products, a Part of SP
Specifications
- Descripción:Disposal carton, benchtop
- Dimensiones:20x20x25 cm (8x8x10")
- Referencia:30617-648
- Nº de proveedor:246530002
Specifications
About this item
Sturdy corrugated cardboard receptacle is supplied with a 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids.
- Sturdy design
- Packaged flat and easy to assemble
- Corrugated cardboard construction
- Supplied with polyethylene bag
When the box is full, lift the flap opening to a closed position and the entire unit is ready for disposal. Printed with eye-catching graphics in four languages (English, German, French, Spanish), it is packaged flat to save space and shipping costs and is quickly assembled. Does not qualify as a DOT container.