ContactUsLinkComponent
Scout® Portable Topload Balance with Square Pan and Touchscreen Display, OHAUS®
Scout® Portable Topload Balance with Square Pan and Touchscreen Display, OHAUS®
Catalog # 10805-292
Supplier:  OHAUS SCALE CORP
cUL Listed
CSA Certified
UL Listed
undefined
Scout® Portable Topload Balance with Square Pan and Touchscreen Display, OHAUS®
Catalog # 10805-292
Supplier:  OHAUS SCALE CORP
Supplier Number:  30253014
cUL Listed
CSA Certified
UL Listed
Specifications
  • Tamaño de la plataforma/la charola:
    140×170 mm (5.5×6.7")
  • Descripción:
    Scout® Model STX2201, Portable Topload Balance with Pan and Touchscreen Display
  • Referencia:
    10805-292
  • Legibilidad:
    0.1 g
  • Capacidad de pesaje:
    2200 g
  • Nº de proveedor:
    30253014
Specifications
About this item
Ideal for laboratory and industrial applications, the OHAUS® Scout® features an easy to navigate color touchscreen display and a slim design which allows stackable storage. Geared for superior performance with fast stabilization time and high resolution weighing results, these portable balances set a new standard for a broad range of weighing applications. Features also include superior overload protection and expanded connectivity options.
  • Informative Color Touchscreen Display
  • Superior Overload Protection and Stackable Storage
Additional standard features include; transportation lock, stainless steel pan, menu and calibration lockout switch, slip-resistant and adjustable feet, leveling bubble, mechanical and software overload/underload protection, stability indicator, auto tare, low battery indicator, auto shut-off, user selectable printing options, and user selectable communication settings.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Totalization, Density Determination, Display Hold, and Mole Weighing.
Note: Model STX421 has a round pan, all other models have a square pan.
Certifications: IEC/EN 61010-1; CAN/CSA C22.2 No. 61010-1; UL Std. No. 61010-1, IEC/EN 61326-1 Class B; FCC Part 15 Class B; Industry Canada ICES-003 Class B.
Ordering information: Includes AC adapter (4 AA batteries not included).