Pitched Blade Propellers, Caframo
Pitched Blade Propeller, with Impeller and Shaft, 6.4 cm (2.5")
Catalog # 52428-754
Supplier: CAFRAMO LTD CA
Número de proveedor #: A166
Jump to:
Product Details & Documents
Caframo 2.5" diameter pitched blade propeller is cast 316L electropolished stainless steel with 5/16" bore
Designed to offer low shear, high top-to-bottom pumping action with high gas (air) injection. Axial flow.
Set screws are included, and attach the impeller to shaft.
Ordering information: This pitched blade propeller can be ordered as an impeller only, or include a 16" stainless steel shaft. Appropriate for use with all Caframo overhead stirrers.
Documents
Caframo Lab Solutions
Documentación del producto
Especificaciones
- Ø:6.4 cm (2.5")
- Description:Pitched Blade Propeller, with Impeller and Shaft