Dropper Bottle, LDPE, 2 oz.
LDPE Dropper Bottle w/ Cap, 2oz
Catalog # 470123-901
Supplier: QORPAK
Número de proveedor #: 501185
Jump to:
Product Details & Documents
Natural LDPE round dropper bottle with attached white cap, 2 oz.
Especificaciones
- Description:LDPE Dropper Bottle w/ Cap, 2oz
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



