Orders LinkContactUsLinkComponent
Ward's® Apatite (Massive)
Ward's® Apatite (Massive)
Supplier:   Avantor
Ward's® Apatite (Massive)
Supplier:  Avantor
Certificates
Ward's® Apatite (Massive)
Ward's® Apatite (Massive)
4 Options Available Below
Product Details & Documents
Blue fluorapatite, pure crystalline, Moh's hardness of 5.
  • Collected in Madagascar
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Specimen Dimensions
    Availability
    Price
  • 470024-886
    Apatite thin section
    Apatite thin section
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,043.40
    $1,043.40
    Each (1UN)
       $1,043.40
  • 470218-154
    Apatite (massive), student specimens
    Apatite (massive), student specimens
    1×1 - 1×1"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $434.70
    $434.70
    Paquete de 10
       $434.70
  • 470218-156
    Apatite (massive), hand specimen
    Apatite (massive), hand specimen
    2×3 - 3×4"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $465.63
    $465.63
    Each (1UN)
       $465.63
  • 470218-158
    Apatite (massive), bulk pack, 1 kg
    Apatite (massive), bulk pack, 1 kg
    Size will vary
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $747.43
    $747.43
    Each (1UN)
       $747.43