General Purpose Manual Syringes, MicroVolume, SGE
Catalog # 60361-068
Supplier: Trajan Scientific and Medical
Specifications
- Ext.Ø:0.47 mm
- Volumen:10 µl
- Descripción:Plunger Protection
- Referencia:60361-068
- Needle Length:50 mm
- Barrel O.D.:6.5 mm
- Needle Gauge:26
- Modelo:10F
- Peso:2 oz.
- Nº de proveedor:002000
- Needle Tip:Bevel
Specifications
About this item
General purpose plunger-in-barrel microvolume syringes
- Plunger Protection incorporates a guide and button reinforcement to minimize the risk of bent plungers
- SuperfleX Flexible Plungers use a super elastic alloy plunger that will not bend or kink
- Guided Plungers have an extended barrel and reinforced plunger that provides additional protection
- Accuracy and reproducibility ±1%
Scale Length of 54.1 mm for 5 µL and 10 µL syringes; 60 mm for 25 µL to 500 µL syringes.
Each plunger is individually fitted for each microsyringe for perfect sealing.