ContactUsLinkComponent
Round-Bottom Three-Neck Flasks, Angled Side Necks, Heavy Wall, Ace Glass Incorporated
Round-Bottom Three-Neck Flasks, Angled Side Necks, Heavy Wall, Ace Glass Incorporated
Catalog # 89056-880
Supplier:  ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
undefined
Round-Bottom Three-Neck Flasks, Angled Side Necks, Heavy Wall, Ace Glass Incorporated
Catalog # 89056-880
Supplier:  ACE GLASS, INC.
Supplier Number:  6948-267
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacidad:
    5 L
  • Descripción:
    Heavy Wall
  • Referencia:
    89056-880
  • Center Neck [ST] Joint:
    45/50
  • Side Neck ST Joints:
    24/40
  • Nº de proveedor:
    6948-267
Specifications
About this item
These round-bottom flasks have angled side necks and [ST] outer joints
The outer joints are tooled with reinforcement rings (full-sized joints only) for added strength and stability. The flare-out directly below the center joint facilitates cleaning of the flask and enables stirring blades to be easily removed. Fabricated from borosilicate glass. Heavy wall flasks feature walls that are approximately 30% heavier than standard wall flasks.