Sterile Centrifuge Tubes, Tarsons
Catalog # 470219-158
Supplier: VWR International
Specifications
- Descripción:Disposable Centrifuge Tubes with Caps, Sterile, 120mm
- Referencia:470219-158
- Nº de proveedor:546021
Specifications
About this item
Conical bottom, polypropylene centrifuge tubes are supplied with cap.
Tubes are sterile and autoclavable. Length: 120 mm. Capacity: 15 ml.