Cryogenic Printer Labels, Brady
Catalog #
Product Details & Documents
Cryogenic labels for tubes, vials, and canes:
Temperatures from –196 to 121 °C (–321 to 250 °F) and hot water baths
B-461 Ultra-thin label with clear tail for tubes.
- General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more
- B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags; working temperatures from –196 to +121 °C (–321 to +250 °F)
- B-595 General purpose label for drawers, shelves, and floor
Chemical and stain resistant labels for slides and cassettes.