Halogen Bulbs, Philips, Bulbtronics©
Bulb, Philips, 19.81 mm Light Center Length (also replacement bulb for microscope models 1130, DXH110BU-F, DXH110BU-Q, H110BG-E, H110BU-B, H110BU-E, H110BU-H, H110BU-PP4, H110BU-PPU, H110TU-H, H110TU-PP4, XH110BG-J, XH110BG-Q, XH110BU-F, XH110BU-J, XH110TG-F XH110TG-Q, XH110TU-F, XH110TU-Q), C6/single coil transverse, 100 h, 31 mm, 9.53 mm, T3, G4/2-pin, 6 V, 20 W, 7388, 3200 K
Catalog # 47738-578
Supplier: BULBTRONICS INC.
Número de proveedor #: 0000986
Jump to:
Product Details & Documents
Various halogen bulbs.
Especificaciones
- Ø:9.53 mm
- Max. temp.:3200 K
- Description:Bulb, Philips, 19.81 mm Light Center Length (also replacement bulb for microscope models 1130, DXH110BU-F, DXH110BU-Q, H110BG-E, H110BU-B, H110BU-E, H110BU-H, H110BU-PP4, H110BU-PPU, H110TU-H, H110TU-PP4, XH110BG-J, XH110BG-Q, XH110BU-F, XH110BU-J, XH110TG-F XH110TG-Q, XH110TU-F, XH110TU-Q)
- Length:31 mm
- Filament:C6/single coil transverse
- Bulb Life:100 h
- Base:G4/2-pin
- Model:T3
- Power:20 W
- Voltage:6 V
- OEM Number:7388