GsBP®-1 Non-Polar GC Columns, 30 - 60 m, GS-Tek
Catalog # 10498-706
Supplier: GENERAL SEPARATION TECHNOLOGIES
Specifications
- Formato de columna:Capillary
- Espesor de la película:0.25 µm
- Ø int.:0.25 mm
- Longitud:30 m
- Temperature limit:-60 to 325/350°C
- Descripción:10498-706
- Referencia:10498-706
- Nº de proveedor:0125-3002
Specifications
About this item
General column for most popular applications, particularly for petroleum and petrochemical industries.
- 100% poly(methyl siloxane) phase, cross-linked and bonded
- Solvent rinse-able and high temperature limit
- Low column bleed, inert and neutral surface, high plate number
- Non-polar, equivalent to USP G1, G2, G38
Ordering information: These columns are suitable replacements for HP-1, DB-1, CP-Sil 5Cb, VF-1, Rtx-1, Rxi-1, SP-1 ZB-1, BP-1.