Orders LinkContactUsLinkComponent
General Purpose Viscosity Standards, Cannon Instruments
General Purpose Viscosity Standards, Cannon Instruments
Certificates
General Purpose Viscosity Standards, Cannon Instruments
14 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These certified standards are useful for calibration and verification procedures.
  • It is suitable for all types of viscometers
  • It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.
Documents
Product Family Options
Información del producto
  • Size
    Viscosity
    Availability
    Price
  • 10021-260
    S600 viscosity standard
    S600 viscosity standard
    500 ml
    (20°C) 1200 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $3,520.88
    $3,520.88
    Each (500ml)
       $3,520.88
  • 10021-118
    Viscosity standard N.4
    Viscosity standard N.4
    500 ml
    (20°C) 0.32 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1ml)
    Each (1ml) - $4,040.08
    $4,040.08
    Each (1ml)
       $4,040.08
  • 47735-104
    Viscosity Standard N10
    Viscosity Standard N10
    500 ml
    (20°C) 21 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,520.88
    $3,520.88
    Each (1UN)
       $3,520.88
  • 10022-026
    Viscosity standard N100
    Viscosity standard N100
    0.5 L
    (20°C) 283 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $3,573.71
    $3,573.71
    Each (500ml)
       $3,573.71
  • 10021-234
    Viscosity standard N250
    Viscosity standard N250
    500 ml
    (20°C) 650 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $3,520.88
    $3,520.88
    Each (500ml)
       $3,520.88
  • 10021-180
    Viscosity standard N26
    Viscosity standard N26
    500 ml
    (20°C) 47 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $3,520.88
    $3,520.88
    Each (500ml)
       $3,520.88
  • 10021-188
    Viscosity standard N35
    Viscosity standard N35
    500 ml
    (20°C) 75 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1ml)
    Each (1ml) - $4,019.69
    $4,019.69
    Each (1ml)
       $4,019.69
  • 10021-242
    Viscosity standard N350
    Viscosity standard N350
    500 ml
    (20°C) 820 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1ml)
    Each (1ml) - $4,019.69
    $4,019.69
    Each (1ml)
       $4,019.69
  • 10021-244
    Viscosity standard N350
    Viscosity standard N350
    1 L
    (20°C) 820 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1ml)
    Each (1ml) - $6,163.94
    $6,163.94
    Each (1ml)
       $6,163.94
  • 10021-270
    Viscosity standard N750
    Viscosity standard N750
    500 ml
    (20°C) 2200 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1ml)
    Each (1ml) - $3,961.03
    $3,961.03
    Each (1ml)
       $3,961.03
  • 10021-170
    Viscosity Standard S20
    Viscosity Standard S20
    500 ml
    (20°C) 37 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1ml)
    Each (1ml) - $3,520.88
    $3,520.88
    Each (1ml)
       $3,520.88
  • 47735-114
    Viscosity Standard S200
    Viscosity Standard S200
    500 ml
    (20°C) 460 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,520.88
    $3,520.88
    Each (1UN)
       $3,520.88
  • 10021-224
    Viscosity standard S200
    Viscosity standard S200
    500 ml
    (20°C) 460 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $3,520.88
    $3,520.88
    Each (500ml)
       $3,520.88
  • 47735-118
    Viscosity Standard S600
    Viscosity Standard S600
    500 ml
    (20°C) 1500 mPa·s
    Discontinued
  • 10021-206
    Viscosity standard S60
    Viscosity standard S60
    500 ml
    (20°C) 170 mPa·s
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $3,520.88
    $3,520.88
    Each (500ml)
       $3,520.88
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 