ContactUsLinkComponent
SPE Vacuum Manifolds, Bonna-Agela Technologies
SPE Vacuum Manifolds, Bonna-Agela Technologies
Catalog # 89213-858
Supplier:  Phenomenex
SPE Vacuum Manifolds, Bonna-Agela Technologies
Catalog # 89213-858
Supplier:  Phenomenex
Supplier Number:  VM12
Specifications
  • Descripción:
    SPE Vacuum Manifold
  • Referencia:
    89213-858
  • No. of Ports:
    12
  • Nº de proveedor:
    VM12
Specifications
About this item
SPE vacuum manifolds are ideal for SPE sample preparation, filtration and elution.
  • Provides consistent extraction and filtration results
  • Solvent resistant and autoclavable lid
  • Adjustable racks will accommodate a variety of sample collection vessels