Orders LinkContactUsLinkComponent
Specific Heat Demonstration Set
Specific Heat Demonstration Set
Supplier:   VWR International
Specific Heat Demonstration Set
Supplier:  VWR International
Certificates
Specific Heat Demonstration Set
Specific Heat Demonstration Set
2 Options Available Below
Product Details & Documents
Visually illustrate the difference in specific heat.
  • Illustrate the different heat capacities of metal
A dramatic way to illustrate the different heat capacities of metal, this slide features a slanted panel with four grooves to hold four different metals. Simply place a sheet of bee's wax in the front of the plane and submerge the metal samples in hot water. When the metals are placed on the grooves simultaneously they will melt and sink through to different levels based on their specific heat. Includes four metals cylinders: aluminum, copper, lead, and tin; support stand, two pieces of bees wax, and instructions.
Product Family Options
Información del producto
  • Availability
    Price
  • 470218-912
    Specific Heat Demonstration Set
    Specific Heat Demonstration Set
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $2,467.83
    $2,467.83
    Kit de 1
       $2,467.83
  • 470218-914
    Wax Paper Replacement, Pack of 2
    Wax Paper Replacement, Pack of 2
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 2
    Paquete de 2 - $437.05
    $437.05
    Paquete de 2
       $437.05