Cartesian Diver Set
Cartesian Diver Set, 1
Catalog # 470218-908
Supplier: VWR International
Jump to:
Product Details & Documents
This diver set expresses the effects of air pressure and buoyancy.
- Topics Include Transmission of Pressure by Liquid, Compressibility of Gases, and Archimedes’ Principle
- Set Also Expresses Effects of Air Pressure and Buoyancy
- Instructions Included
This apparatus is ideal for showing the transmission of pressure by a liquid, the compressibility of gases, and Archimedes’ Principle.
Instructions are included.
Ordering information: This kit includes a Cartesian diver, a 25 x 4 cm dia. glass hydrometer cylinder, a rubber diaphragm, and instructions.
Specifications
- Description:Cartesian Diver Set
- No. of Lab Groups:1