Orders LinkContactUsLinkComponent
Examination Gloves, Microflex®, Ansell
Examination Gloves, Microflex®, Ansell
Supplier:   Ansell Healthcare
Examination Gloves, Microflex®, Ansell
Supplier:  Ansell Healthcare
Certificates
Examination Gloves, Microflex®, Ansell
3 Options Available Below
Product Details & Documents
This thicker version of the popular Microflex® Cobalt® X N21 glove offers fit and feel with the extra protection of a thicker glove.
  • Fully textured to provide grip in any condition
  • Non sterile
  • Ambidextrous
  • Gloves are blue and feature beaded cuffs
For use in life science, electronics, and food processing industries for analytical testing/measurements; assembly and inspection; biotechnologies; cleaning; food processing; handling small parts; laboratory analysis; laboratory testing; light duty maintenance and clean up; light assembly tasks; maintaining equipment and instruments; pharmaceutical manufacturing; preparing pharmaceutical products; sanitation and dish washing; and as an under glove for meat/fish processing.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 10770-436
  • 10770-460
  • 10770-462
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 3.9 mil
  • 3.9 mil
  • 3.9 mil
  • Finger thickness:
  • 5.5 mil
  • 5.5 mil
  • 5.5 mil
  • Palm thickness:
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
Especificaciones
Product Family Options
Información del producto
  • Glove size
    Glove Color
    Length
    Availability
    Price
  • S
    Blue
    24 cm (9⅜")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $347.00
    $347.00
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $419.90
    $419.90
    Paquete de 100
       $347.00
    Case de 10 (100UN)
       $419.90
    Supplier Number: N211
    Especificaciones:
    More
    Specifications
    Cat. No.
    10770-436
    Ambidextrous / hand-specific
    Ambidextrous
    Cuff style
    Beaded
    Cuff thickness
    3.9 mil
    Finger thickness
    5.5 mil
    Palm thickness
    4.7 mil
  • M
    Blue
    24 cm (9⅜")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $347.00
    $347.00
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $419.90
    $419.90
    Paquete de 100
       $347.00
    Case de 10 (100UN)
       $419.90
    Supplier Number: N212
    Especificaciones:
    More
    Specifications
    Cat. No.
    10770-460
    Ambidextrous / hand-specific
    Ambidextrous
    Cuff style
    Beaded
    Cuff thickness
    3.9 mil
    Finger thickness
    5.5 mil
    Palm thickness
    4.7 mil
  • L
    Blue
    24 cm (9⅜")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $347.00
    $347.00
    Log in Para disponibilidad
    Case de 10 (100UN)
    Case de 10 (100UN) - $419.90
    $419.90
    Paquete de 100
       $347.00
    Case de 10 (100UN)
       $419.90
    Supplier Number: N213
    Especificaciones:
    More
    Specifications
    Cat. No.
    10770-462
    Ambidextrous / hand-specific
    Ambidextrous
    Cuff style
    Beaded
    Cuff thickness
    3.9 mil
    Finger thickness
    5.5 mil
    Palm thickness
    4.7 mil