Crucibles with Lids, Quartz, Chemglass
89029-684
Small Business Enterprise
- Capacidad:40 ml
- Descripción:Crucible
- Altura:45 mm
- Referencia:89029-684
- Diámetro de la boca:57 mm
- Diámetro del fondo:20 mm
- Nº de proveedor:CGQ-4000-03
Crucibles are suitable for use in temperatures up to 1000°C, which is perfect for use in muffle furnace ashing procedures.
- Made of quartz
- Withstands temperatures up to 1000°C
Quartz material ensures sample purity for further analysis.