Orders LinkContactUsLinkComponent
Freeze-Thaw Chambers
Freeze-Thaw Chambers
   Opciones sostenibles disponibles
Freeze-Thaw Chambers
   Opciones sostenibles disponibles
Certificates
Freeze-Thaw Chambers
2 Options Available Below
Product Details & Documents
For precise environmental control, modelling short-term low temperature conditions. This broad range encompasses nearly every ambient environment on earth. Automatic, programmable, and accurate cycling between setpoints enables continuous simulation of harsh freezing and thawing conditions for rapid and repeatable materials testing.
  • Wide −25 to +70 °C temperature range for full simulation of freeze/thaw cycles and temperature excursions
  • Easy cycle programming through large-format color touchscreen
  • Feature up to 5 user-defined programs, with 50 individual steps, up to 255 loops, and a ramp based on any time or rate
  • Available humidity control
Caron's chambers are built for many types of freeze/thaw studies, including pharmaceutical drug excursion testing, concrete and asphalt testing, coating and adhesive testing, outdoor weathering tests, and food, beverage, and packaging testing.
Multiple optional accessories are also available to allow you to customize the chamber to match your specific needs.
Certifications: CE.
Ordering information: Accessories available separately.
Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product. PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 10216-800
  • 10216-804
  • 85022-690
  • Temperature range:
  • −25…+70 °C
  • −25…+70 °C
  • −25…+70 °C
  • Environmentally Preferable:
  • Sustainable Packaging|Energy Efficient
  • Sustainable Packaging|Energy Efficient
  • Temperature control:
  • ±0.1 °C
  • ±0.1 °C
  • ±0.1 °C
  • Temperature sensor:
  • 3-wire RTD
  • 3-wire RTD
  • 3-wire RTD
  • Temperature uniformity:
  • ±0.3 °C
  • ±0.3 °C
  • ±0.3 °C
Especificaciones
Product Family Options
Información del producto
  • Electrical
    Interior Dimensions
    Humidity Range
    Volume
    Shipping Weight
    Availability
    Price
  • 10216-800
    Freeze/thaw chamber, model 7900-25-2 with capacitive humidity sensor
    Freeze/thaw chamber, model 7900-25-2 with capacitive humidity sensor
    208/230 V, 60 Hz,12 A
    81W×68.6D×133.9H cm (32×27×52.7")
    20 - 98% RH
    708 L (25 cu. ft.)
    356 kg (785 lbs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $700,203.09
    $700,203.09
    Each (1UN)
       $700,203.09
    Supplier Number: 7900-25-2
    Especificaciones:
    Product available on GSA Advantage®
    Small Business Enterprise
      Sustainable Packaging|Energy Efficient
    More
    Specifications
    Cat. No.
    10216-800
    Temperature range
    −25…+70 °C
    Environmentally Preferable
    Sustainable Packaging|Energy Efficient
    Temperature control
    ±0.1 °C
    Temperature sensor
    3-wire RTD
    Temperature uniformity
    ±0.3 °C
    Product available on GSA Advantage®
    Small Business Enterprise
      Sustainable Packaging|Energy Efficient
  • 10216-804
    Freeze/thaw chamber, model 7900-33-2 with capacitive humidity sensor
    Freeze/thaw chamber, model 7900-33-2 with capacitive humidity sensor
    208/230 V, 60 Hz,12 A
    81W×68.6D×166.9H cm (32×27×65.7")
    20 - 98% RH
    934 L (33 cu. ft.)
    399 kg (880 lbs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $963,398.61
    $963,398.61
    Each (1UN)
       $963,398.61
    Supplier Number: 7900-33-2
    Especificaciones:
    Product available on GSA Advantage®
    Small Business Enterprise
      Sustainable Packaging|Energy Efficient
    More
    Specifications
    Cat. No.
    10216-804
    Temperature range
    −25…+70 °C
    Environmentally Preferable
    Sustainable Packaging|Energy Efficient
    Temperature control
    ±0.1 °C
    Temperature sensor
    3-wire RTD
    Temperature uniformity
    ±0.3 °C
    Product available on GSA Advantage®
    Small Business Enterprise
      Sustainable Packaging|Energy Efficient
  • 85022-690
    Freeze/thaw chamber, model 7900-33-2 with capacitive humidity sensor
    Freeze/thaw chamber, model 7900-33-2 with capacitive humidity sensor
    208/230 V, 60 Hz, 12 A
    81W×68.6D×166.9H cm (32×27×65.7")
    20 - 98% RH
    934 L (33 cu. ft.)
    399 kg (880 lbs.)
    Discontinued
    Supplier Number: -
    Especificaciones:
    More
    Specifications
    Cat. No.
    85022-690
    Temperature range
    −25…+70 °C
    Environmentally Preferable
    Temperature control
    ±0.1 °C
    Temperature sensor
    3-wire RTD
    Temperature uniformity
    ±0.3 °C