Amber Plain Disposable Borosilicate Glass Tubes, Kimble Chase
Catálogo # 10799-210
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Especificaciones
- Ext.Ø:16 mm
- Volumen:15 ml (overflow capacity)
- Longitud:100 mm
- Referencia:10799-210
- Nº de proveedor:73500-16100A
Especificaciones
Acerca de este artículo
Premium quality tubes provide superior protection for light sensitive applications.
Tubes are manufactured from amber 51 expansion borosilicate glass.
Embalaje: Packed in convenient trays that are shrink-wrapped in plastic film.